Merck
  • The "Mutated in Colorectal Cancer" Protein Is a Novel Target of the UV-Induced DNA Damage Checkpoint.
The "Mutated in Colorectal Cancer" Protein Is a Novel Target of the UV-Induced DNA Damage Checkpoint.
Genes & cancer (2011-07-23)
Laurent Pangon, Nicholas D Sigglekow, Mark Larance, Sam Al-Sohaily, Dessislava N Mladenova, Christina I Selinger, Elizabeth A Musgrove, Maija R J Kohonen-Corish
ABSTRACT
MCC is a potential tumor suppressor gene, which is silenced by promoter hypermethylation in a subset of colorectal cancers. However, its functions have remained poorly understood. In the present study, we describe a novel function of MCC in the DNA damage response. Several novel phosphorylation sites were identified by mass spectrometry, including 2 highly conserved ATM/ATR consensus sites at serine 118 and serine 120. In addition, exposure to ultraviolet radiation (UV), but not phleomycin, caused PI3K-dependent phosphorylation of MCC and its nuclear localization. Re-expression of MCC in HCT15 colorectal cancer cells led to a G2/M arrest, and MCC knockdown impaired the induction of a G2/M arrest following UV radiation. Finally, mutation of S118/120 to alanine did not affect MCC nuclear shuttling following UV but did impair MCC G2/M checkpoint activity. Thus, these results suggest that MCC is a novel target of the DNA damage checkpoint and that MCC is required for the complete cell cycle arrest in the G2/M phase in response to UV.
Sigma-Aldrich
Anti-phospho-Histone H3 (pSer10) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)