Merck
  • Study of an aqueous lithium chloride desiccant system: air dehumidification and desiccant regeneration
Study of an aqueous lithium chloride desiccant system: air dehumidification and desiccant regeneration
Solar Energy (2002)
Fumo, Nelson, and D. Y. Goswami.