- Catalytic asymmetric synthesis promoted by a chiral zirconate: Highly enantioselective allylation of aldehydes
Catalytic asymmetric synthesis promoted by a chiral zirconate: Highly enantioselective allylation of aldehydes
Tetrahedron Letters (1995)
Bedeschi P, et al.