Merck
  • Biological effects of propionic acid in humans; metabolism, potential applications and underlying mechanisms
Biological effects of propionic acid in humans; metabolism, potential applications and underlying mechanisms
Molecular and Cellular Biology (2010)
Sa'ad, H., et al.