- Comparison of coliforms and coliphages as tools for assessment of viral contamination in river water.
Comparison of coliforms and coliphages as tools for assessment of viral contamination in river water.
Applied and Environmental Microbiology (2004)
Skraber S et al