Merck
  • Polymorphic exact tandem repeat A (PETRA): a newly defined lineage of mycobacterium tuberculosis in israel originating predominantly in Sub-Saharan Africa.
Polymorphic exact tandem repeat A (PETRA): a newly defined lineage of mycobacterium tuberculosis in israel originating predominantly in Sub-Saharan Africa.
Journal of Clinical Microbiology (2009)
Freidlin PJ et al