- Presence of viral genomes in mineral water: a sufficient condition to assume infectious risk?
Presence of viral genomes in mineral water: a sufficient condition to assume infectious risk?
Applied and Environmental Microbiology (2003)
Gassilloud B et al