- Crystal structure of core streptavidin determined from multiwavelength anomalous diffraction of synchrotron radiation.
Crystal structure of core streptavidin determined from multiwavelength anomalous diffraction of synchrotron radiation.
Proceedings of the National Academy of Sciences of the USA (1989)
Hendrickson WA et al