- Role of Mitochondrial Metabolism in the Control of
Early Lineage Progression and Aging Phenotypes in
Adult Hippocampal Neurogenesis
Role of Mitochondrial Metabolism in the Control of<BR/> Early Lineage Progression and Aging Phenotypes in<BR/> Adult Hippocampal Neurogenesis
Neuron (2016)
Ruth Beckervordersandforth