Merck
  • Inulin-type fructan degradation capacity of Clostridium cluster IV and XIVa butyrate-producing colon bacteria and their associated metabolic outcomes.
Inulin-type fructan degradation capacity of Clostridium cluster IV and XIVa butyrate-producing colon bacteria and their associated metabolic outcomes.
Beneficial microbes (2017-05-27)
F Moens, L De Vuyst
ABSTRACT
Four selected butyrate-producing colon bacterial strains belonging to Clostridium cluster IV (Butyricicoccus pullicaecorum DSM 23266
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG