Merck
  • Effects of intrauterine infusion of Escherichia coli lipopolysaccharide on uterine health, resolution of purulent vaginal discharge, and reproductive performance of lactating dairy cows.
Effects of intrauterine infusion of Escherichia coli lipopolysaccharide on uterine health, resolution of purulent vaginal discharge, and reproductive performance of lactating dairy cows.
Journal of dairy science (2017-04-25)
João G N Moraes, Paula R B Silva, Luís G D Mendonça, Alexandre A Scanavez, Joseane C C Silva, Ricardo C Chebel
ABSTRACT
The objectives of the current experiment were to evaluate the effects of intrauterine infusion of Escherichia coli lipopolysaccharide (LPS) in cows diagnosed with purulent vaginal discharge (PVD) on intrauterine cell population, resolution of PVD, uterine health, and reproductive performance. Jersey cows (n = 3,084) were examined using the Metricheck device to diagnose PVD at 35 ± 6 d postpartum. Purulent vaginal discharge was defined as the presence of purulent (≥50% pus) discharge detectable in the vagina. Of the 310 cows positive for PVD, 267 cows were enrolled in the current experiment. To ensure proper timing of treatment and collection of samples, only 9 PVD-positive cows were treated per day. Selected cows were balanced at 35 ± 6 d postpartum for lactation number, body condition score, and milk yield and were randomly assigned to receive an intrauterine infusion of 20 mL of phosphate-buffered saline (PBS; control, n = 87), 20 mL of PBS with 150 µg LPS (LPS150, n = 91), or 20 mL of PBS with 300 µg of LPS (LPS300, n = 89). Uterine cytology was performed immediately before treatment and 1, 2, and 7 d after treatment to evaluate the effect of LPS treatment on intrauterine cell population. Cows were examined with the Metricheck device at 7 and 28 d after treatment to evaluate the effects of treatment on resolution of PVD. Reproductive status was recorded up to 200 d postpartum. Cows diagnosed with PVD had greater incidence of twinning, dystocia, retained placenta, and metritis after calving than cows without PVD. Count of polymorphonuclear leukocytes (PMNL) in uterine cytology 1, 2, and 7 d after intrauterine infusion was not statistically different among treatments. From d 0 to 1, however, PMNL count in uterine cytology of PBS cows increased by 5%, whereas the PMNL count in uterine cytology of LPS150 and LPS300 cows increased by 54 and 48%, respectively. Treatment did not affect the likelihood of cows being diagnosed with PVD 7 and 28 d after intrauterine infusion. Cows without PVD and LPS150 cows were more likely to be pregnant after the first postpartum AI than PBS cows. After the second postpartum AI, cows without PVD were more likely to be pregnant than PBS and LPS300 cows. Hazard of pregnancy up to 200 d postpartum was decreased for PBS and LPS300 cows compared with cows without PVD, and it tended to be decreased for LPS150 cows compared with cows without PVD. Intrauterine treatment with 150 µg of E. coli LPS of cows diagnosed with PVD improved likelihood of pregnancy after the first postpartum AI, but further research is needed to elucidate the mechanism by which LPS treatment improved fertility.
Product Description
Supelco
Ceftiofur hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction