Merck
  • Genetic requirement for Mycl and efficacy of RNA Pol I inhibition in mouse models of small cell lung cancer.
Genetic requirement for Mycl and efficacy of RNA Pol I inhibition in mouse models of small cell lung cancer.
Genes & development (2016-06-15)
Dong-Wook Kim, Nan Wu, Young-Chul Kim, Pei Feng Cheng, Ryan Basom, Dongkyoon Kim, Colin T Dunn, Anastasia Y Lee, Keebeom Kim, Chang Sup Lee, Andrew Singh, Adi F Gazdar, Chris R Harris, Robert N Eisenman, Kwon-Sik Park, David MacPherson
ABSTRACT
Small cell lung cancer (SCLC) is a devastating neuroendocrine carcinoma. MYCL (L-Myc) is frequently amplified in human SCLC, but its roles in SCLC progression are poorly understood. We isolated preneoplastic neuroendocrine cells from a mouse model of SCLC and found that ectopic expression of L-Myc, c-Myc, or N-Myc conferred tumor-forming capacity. We focused on L-Myc, which promoted pre-rRNA synthesis and transcriptional programs associated with ribosomal biogenesis. Deletion of Mycl in two genetically engineered models of SCLC resulted in strong suppression of SCLC. The high degree of suppression suggested that L-Myc may constitute a therapeutic target for a broad subset of SCLC. We then used an RNA polymerase I inhibitor to target rRNA synthesis in an autochthonous Rb/p53-deleted mouse SCLC model and found significant tumor inhibition. These data reveal that activation of RNA polymerase I by L-Myc and other MYC family proteins provides an axis of vulnerability for this recalcitrant cancer.
Sigma-Aldrich
Anti-UCHL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse