Merck
  • Stimulation of μ-opioid receptors dilates retinal arterioles by neuronal nitric oxide synthase-derived nitric oxide in rats.
Stimulation of μ-opioid receptors dilates retinal arterioles by neuronal nitric oxide synthase-derived nitric oxide in rats.
European journal of pharmacology (2017-03-28)
Eriko Someya, Asami Mori, Kenji Sakamoto, Kunio Ishii, Tsutomu Nakahara
ABSTRACT
Opioids contribute to the regulation of cerebral vascular tone. The purpose of this study was to examine the effects of herkinorin, a non-opioid μ-opioid receptor agonist derived from salvinorin A, on blood vessels in the rat retina and to investigate the mechanism underlying the herkinorin-induced retinal vasodilatory response. Ocular fundus images were captured using an original high-resolution digital fundus camera in vivo. The retinal vascular responses were evaluated by measuring the diameter of retinal arterioles in the fundus images. Both systemic blood pressure and heart rate were continuously recorded. Herkinorin increased the retinal arteriolar diameter without significantly changing mean blood pressure and heart rate. The retinal vasodilator response to herkinorin was almost completely prevented following treatment with naloxone, a nonselective opioid receptor antagonist and H-D-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
CTOP, ≥97% (HPLC)