Merck
  • Fuzheng Qingjie Granules Inhibit Growth of Hepatoma Cells via Inducing Mitochondria-Mediated Apoptosis and Enhancing Immune Function.
Fuzheng Qingjie Granules Inhibit Growth of Hepatoma Cells via Inducing Mitochondria-Mediated Apoptosis and Enhancing Immune Function.
Integrative cancer therapies (2016-06-24)
Xuzheng Chen, Zhiyun Cao, Youquan Zhang, Jinnong Li, Suqing Wang, Jian Du, Lianming Liao
ABSTRACT
Fuzheng Qingjie (FZQJ) granules, a compound Chinese medicine, have been used as an adjuvant therapy for alimentary tract cancers. However, the underlying anticancer mechanisms are still not well understood. In the present study, HepG2 cells were treated with FZQJ-containing serum. Cell proliferation was evaluated using MTT assay. Apoptosis was analyzed using a flow cytometer. Cell ultrastructure was observed under a transmission electron microscope. The mitochondrial membrane potential (Δψ) was examined with JC-1 dye. In H22 tumor-bearing mice, CD4
Sigma-Aldrich
JC-1, powder or solid (Crystals)