Merck
  • Activation of Wnt signaling reduces high-glucose mediated damages on skin fibroblast cells.
Activation of Wnt signaling reduces high-glucose mediated damages on skin fibroblast cells.
Iranian journal of basic medical sciences (2017-11-01)
Youpei Wang, Xiang Zheng, Qing Wang, Meiqin Zheng, Lingxia Pang
ABSTRACT
High-glucose (HG) stress, a mimic of diabetes mellitus (DM) in culture cells, alters expression of a large number of genes including Wnt and NF-κB signaling-related genes; however, the role of Wnt signaling during HG-mediated fibroblast damage and the relationship between Wnt and NF-κB signaling have not been understood. In this study, we aimed to investigate the ffects of Wnt signaling on HG-mediated damages. Wnt3a was treated to HG-stressed human primary foreskin fibroblasts and the levels of Wnt signaling markers and cell proliferation were monitored. In addition, Wnt3a and NF-κB signaling inhibitor were assisted to analyze the relationship between two pathways. The results indicated that HG treatment repressed β-catenin level, and Wnt3a treatment increased the levels of β-catenin and HG positively inhibits Wnt signaling, and signaling activation via supplementation of Wnt3a rescued the defect caused by HG. NF-κB signaling negatively regulates accumulation of β-catenin, but Wnt signaling has no effects on IκBα activation.
Sigma-Aldrich
D-Glutamine, ≥98% (HPLC)