Merck
  • Unveiling Cerebral Leishmaniasis: parasites and brain inflammation in Leishmania donovani infected mice.
Unveiling Cerebral Leishmaniasis: parasites and brain inflammation in Leishmania donovani infected mice.
Scientific reports (2017-08-18)
Guilherme D Melo, Sophie Goyard, Laurence Fiette, Alexandre Boissonnas, Christophe Combadiere, Gisele F Machado, Paola Minoprio, Thierry Lang
ABSTRACT
Visceral leishmaniasis (VL) is a systemic disease with multifaceted clinical manifestations, including neurological signs, however, the involvement of the nervous system during VL is underestimated. Accordingly, we investigated both brain infection and inflammation in a mouse model of VL. Using bioluminescent Leishmania donovani and real-time 2D-3D imaging tools, we strikingly detected live parasites in the brain, where we observed a compartmentalized dual-phased inflammation pattern: an early phase during the first two weeks post-infection, with the prompt arrival of neutrophils and Ly6C
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)
Pricing and availability is not currently available.