Merck
  • Spatial and spectral resolution of carbonaceous material from hematite (α-Fe
Spatial and spectral resolution of carbonaceous material from hematite (α-Fe
The Analyst (2017-07-06)
Joseph P Smith, Frank C Smith, Karl S Booksh
ABSTRACT
The search for evidence of extant or past life on Mars is a primary objective of both the upcoming Mars 2020 rover (NASA) and ExoMars 2020 rover (ESA/Roscosmos) missions. This search will involve the detection and identification of organic molecules and/or carbonaceous material within the Martian surface environment. For the first time on a mission to Mars, the scientific payload for each rover will include a Raman spectrometer, an instrument well-suited for this search. Hematite (α-Fe
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)