- Nitrate transport and not photoinhibition limits growth of the freshwater Cyanobacterium synechococcus species PCC 6301 at low temperature.
Nitrate transport and not photoinhibition limits growth of the freshwater Cyanobacterium synechococcus species PCC 6301 at low temperature.
Plant Physiology (1999)
Sakamoto T and Bryant DA