Merck
  • Engineered Assimilation of Exogenous and Endogenous Formate in Escherichia coli.
Engineered Assimilation of Exogenous and Endogenous Formate in Escherichia coli.
ACS synthetic biology (2017-05-31)
Oren Yishai, Leander Goldbach, Hezi Tenenboim, Steffen N Lindner, Arren Bar-Even
ABSTRACT
Decoupling biorefineries from land use and agriculture is a major challenge. As formate can be produced from various sources, e.g., electrochemical reduction of CO
Sigma-Aldrich
D-Glucose-3-13C, 99 atom % 13C, 99% (CP)
Pricing and availability is not currently available.
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Pricing and availability is not currently available.