Merck
  • Sex differences in Alzheimer risk: Brain imaging of endocrine vs chronologic aging.
Sex differences in Alzheimer risk: Brain imaging of endocrine vs chronologic aging.
Neurology (2017-09-01)
Lisa Mosconi, Valentina Berti, Crystal Quinn, Pauline McHugh, Gabriella Petrongolo, Isabella Varsavsky, Ricardo S Osorio, Alberto Pupi, Shankar Vallabhajosula, Richard S Isaacson, Mony J de Leon, Roberta Diaz Brinton
ABSTRACT
This observational multimodality brain imaging study investigates emergence of endophenotypes of late-onset Alzheimer disease (AD) risk during endocrine transition states in a cohort of clinically and cognitively normal women and age-matched men. Forty-two 40- to 60-year-old cognitively normal women (15 asymptomatic perimenopausal by age [CNT], 13 perimenopausal [PERI], and 14 postmenopausal [MENO]) and 18 age- and education-matched men were examined. All patients had volumetric MRI, As expected, the MENO group was older than the PERI and CNT groups. Otherwise, groups were comparable on clinical and neuropsychological measures and Multimodality brain imaging indicates sex differences in development of the AD endophenotype, suggesting that the preclinical AD phase is early in the female aging process and coincides with the endocrine transition of perimenopause. These data indicate that the optimal window of opportunity for therapeutic intervention in women is early in the endocrine aging process.
Sigma-Aldrich
PiB, ≥98% (HPLC)