Merck
  • Effects of water pH on the uptake and elimination of the piscicide, 3-trifluoromethyl-4-nitrophenol (TFM), by larval sea lamprey.
Effects of water pH on the uptake and elimination of the piscicide, 3-trifluoromethyl-4-nitrophenol (TFM), by larval sea lamprey.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2017-06-08)
Benjamin L Hlina, Laura R Tessier, Michael P Wilkie
ABSTRACT
Invasive sea lamprey (Petromyzon marinus) populations in the Great Lakes are controlled by applying the piscicide, 3-trifluoromethyl-4-nitrophenol (TFM), to infested streams with larval sea lamprey (ammocoetes). While treatment mortality is >90%, surviving lamprey, called residuals, can undermine control efforts. A key determinant of TFM effectiveness is water pH, which can fluctuate daily and seasonally in surface waters. The objectives of this research were to evaluate the influence of pH on the uptake, elimination, and accumulation of TFM by larval sea lamprey using radio-labeled TFM (
Sigma-Aldrich
4-Nitro-3-(trifluoromethyl)phenol, 99% (GC)