Merck
  • Obesity-induced vascular inflammation involves elevated arginase activity.
Obesity-induced vascular inflammation involves elevated arginase activity.
American journal of physiology. Regulatory, integrative and comparative physiology (2017-08-25)
Lin Yao, Anil Bhatta, Zhimin Xu, Jijun Chen, Haroldo A Toque, Yongjun Chen, Yimin Xu, Zsolt Bagi, Rudolf Lucas, Yuqing Huo, Ruth B Caldwell, R William Caldwell
ABSTRACT
Obesity-induced vascular dysfunction involves pathological remodeling of the visceral adipose tissue (VAT) and increased inflammation. Our previous studies showed that arginase 1 (A1) in endothelial cells (ECs) is critically involved in obesity-induced vascular dysfunction. We tested the hypothesis that EC-A1 activity also drives obesity-related VAT remodeling and inflammation. Our studies utilized wild-type and EC-A1 knockout (KO) mice made obese by high-fat/high-sucrose (HFHS) diet. HFHS diet induced increases in body weight, fasting blood glucose, and VAT expansion. This was accompanied by increased arginase activity and A1 expression in vascular ECs and increased expression of tumor necrosis factor-α (TNF-α), monocyte chemoattractant protein-1 (MCP-1), interleukin-10 (IL-10), vascular cell adhesion molecule-1 (VCAM-1), and intercellular adhesion molecule-1 (ICAM-1) mRNA and protein in both VAT and ECs. HFHS also markedly increased circulating inflammatory monocytes and VAT infiltration by inflammatory macrophages, while reducing reparative macrophages. Additionally, adipocyte size and fibrosis increased and capillary density decreased in VAT. These effects of HFHS, except for weight gain and hyperglycemia, were prevented or reduced in mice lacking EC-A1 or treated with the arginase inhibitor 2-(
Sigma-Aldrich
Sodium palmitate, ≥98.5%