Merck
  • Quantitative determination of carfilzomib in mouse plasma by liquid chromatography-tandem mass spectrometry and its application to a pharmacokinetic study.
Quantitative determination of carfilzomib in mouse plasma by liquid chromatography-tandem mass spectrometry and its application to a pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2017-09-18)
Jee Sun Min, Jiseon Kim, Jung Ho Kim, Doyun Kim, Yu Fen Zheng, Ji Eun Park, Wooin Lee, Soo Kyung Bae
ABSTRACT
A highly sensitive and rapid LC-MS/MS method was developed and validated to determine the levels of carfilzomib in mice plasma by using chlorpropamide as an internal standard. Carfilzomib and chlorpropamide were extracted from 5 μL of plasma after protein precipitation with acetonitrile. Chromatographic separation was performed on Phenomenex Luna C
Product Description
Supelco
Chlorpropamide, analytical standard, ≥97%