Merck
  • Proteomic Profiling of Liver and Plasma in Chronic Ethanol Feeding Model of Hepatic Alcohol Dehydrogenase-Deficient Deer Mice.
Proteomic Profiling of Liver and Plasma in Chronic Ethanol Feeding Model of Hepatic Alcohol Dehydrogenase-Deficient Deer Mice.
Alcoholism, clinical and experimental research (2017-08-10)
Kamlesh K Bhopale, Samir M Amer, Lata Kaphalia, Kizhake V Soman, John E Wiktorowicz, Ghulam A Shakeel Ansari, Bhupendra S Kaphalia
ABSTRACT
Chronic alcohol abuse, a major risk factor for such diseases as hepatitis and cirrhosis, impairs hepatic alcohol dehydrogenase (ADH; key ethanol [EtOH]-metabolizing enzyme). Therefore, differentially altered hepatic and plasma proteomes were identified in chronic EtOH feeding model of hepatic ADH-deficient (ADH- ) deer mice to understand the metabolic basis of alcoholic liver disease (ALD). ADH- deer mice were fed 3.5 g% EtOH via Lieber-DeCarli liquid diet daily for 3 months and histology of the liver assessed. Liver and plasma proteins were separated by 2-dimensional gel electrophoresis. The proteins differentially expressed were identified by matrix-assisted laser desorption ionization-time of flight mass spectrometry. Histology of the liver showed panlobular steatosis and infiltration of T lymphocytes. Using the criteria of ≥1.5 for fold change (p-value ≤0.05) with expectation value (E ≤10-3 ) and protein score (≥64), 18 proteins in the livers and 5 in the plasma of EtOH-fed mice were differentially expressed and identified. Prolyl 4-hydroxylase, cytochrome b-5, endo A cytokeratin, ATP synthase, heat-shock 70 kD proteins, enoyl CoA hydratase, stress-70 protein, peroxiredoxin 1, and ornithine carbamoyl transferase were up-regulated in the livers. However, carbonic anhydrase 3, mitochondrial ATP synthase, aldolase 2, actin γ, laminin receptor, and carbamoyl phosphate synthase were down-regulated. Contrary to the increased expression of creatine kinase M-type, a decreased expression of serine protease inhibitor A3A precursor, sulfated glycoprotein-2 (clusterin), and apolipoprotein E isoforms were found in the plasma of EtOH group. Chronic EtOH feeding in ADH- deer mice causes steatosis and infiltration of T lymphocytes in the livers along with increased expression of proteins involved in endoplasmic reticulum (ER) stress, fibrosis, fatty acid β oxidation and biogenesis, and decreased expression of proteins involved in ATP synthesis, carbohydrate metabolism, in cell regulation and architecture. Reduced expression of various carrier proteins as found in the plasma of EtOH group has a biomarker potential.
Product Description
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Hsp70 Mouse mAb (C92F3A-5), liquid, clone C92F3A-5, Calbiochem®
Pricing and availability is not currently available.
Millipore
ProteoExtract® Protein Precipitation Kit
Pricing and availability is not currently available.