Merck
Epidemiology of Diarrheal Illness in Pediatric Oncology Patients.
Journal of the Pediatric Infectious Diseases Society (2016-09-01)
Mohammad N Mhaissen, Alicia Rodriguez, Zhengming Gu, Haiqing Zhu, Li Tang, Yilun Sun, Stacey T Schultz-Cherry, Randall T Hayden, Elisabeth E Adderson
ABSTRACT
Diarrhea is common in children with cancer, but this has not been systematically studied to date. Remnant stool samples collected between January 2010 and June 2011 from pediatric oncology patients with diarrhea were tested for bacterial, viral, and parasitic enteropathogens using a combination of standard-of-care (SOC) diagnostic tests, including broad-range, real-time polymerase chain reaction (PCR) assays for adenoviruses, astroviruses, and sapoviruses and 2 commercially available multiplexed PCR assays. Corresponding demographic and clinical data were abstracted from patients' medical records. One hundred fourteen episodes of diarrhea in 93 patients (median age, 3.7 years; range, 0.2-18.8) were included in the study. No patients died, but morbidity was significant. A total of 158 potential pathogens were detected in 114 diarrhea episodes, with >1 organism in one third of these; the most common were Clostridium difficile, noroviruses, adenoviruses, and astroviruses. Clostridium difficile, in combination with norovirus or adenovirus, was most common when >1 pathogen was detected. When both studies were obtained, SOC and broadly multiplexed PCR tests were concordant in 64 episodes (56%). Forty-five pathogens (28%) were identified retrospectively by broadly multiplexed PCR assays only. A total of 19 (13%) were detected by SOC real-time PCR assays but not by either commercially available multiplexed PCR assay. Most pediatric oncology patients in this study had 1 or more potential infectious causes for their diarrhea. Additional studies are warranted to understand the natural history of gastroenteritis in this patient population. Although broadly multiplexed PCR assays offer some advantages over conventional testing, there may be disadvantages to their use for the diagnosis of infectious gastroenteritis that are unique to pediatric oncology patients.
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)