Merck
  • Single-walled carbon nanotube: One specific inhibitor of cancer stem cells in osteosarcoma upon downregulation of the TGFβ1 signaling.
Single-walled carbon nanotube: One specific inhibitor of cancer stem cells in osteosarcoma upon downregulation of the TGFβ1 signaling.
Biomaterials (2017-10-11)
Yanyan Miao, Haixia Zhang, Yubin Pan, Jian Ren, Miaoman Ye, Fangfang Xia, Rui Huang, Zhuoheng Lin, Shuai Jiang, Ya Zhang, Zhou Songyang, Yan Zhang
ABSTRACT
Cancer stem cells (CSCs) are believed to have a critical role in tumorigenesis, metastasis, therapeutic resistance or recurrence. Therefore, strategies designed to specifically target and eliminate CSCs have become one of the most promising and desirable ways for tumor treatment. Osteosarcoma stem cells (OSCs), the CSCs in osteosarcoma (OS), are critically associated with OS progression. Here, we show that single-walled carbon nanotubes (SWCNTs), including unmodified SWCNT (SWCNT-Raw) and SWCNT-COOH, have the ability to specifically inhibit the process of TGFβ1-induced OS cells dedifferentiation, prevent the stem cell phenotypes acquisition in OS cells and reduce the OSC viability under conditions which mimic the OS microenvironment. Concurrently, SWCNT treatment significantly down-regulates the expression of OSC markers in OS, and markedly reduces the tumor microvessel density and tumor growth. Furthermore, we found that SWCNT could suppress the TGFβ1-induced activation of TGFβ type I receptor and downstream signaling, which are key for the OSC formation and maintenance. Our results reveal an unexpected function of SWCNT in negative modulation of OSCs, and provide significant implications for the potential CSCs-targeted therapeutic applications of SWCNT.
Product Description
SAFC
Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12, powder, with 3151 mg/L dextrose, with 2.5 mM L-glutamine, with 55 mg/L sodium pyruvate, without sodium bicarbonate
Pricing and availability is not currently available.