Merck
  • The effect of bilirubin on lipid peroxidation and antioxidant enzymes in cumene hydroperoxide-treated erythrocytes.
The effect of bilirubin on lipid peroxidation and antioxidant enzymes in cumene hydroperoxide-treated erythrocytes.
International journal of clinical & laboratory research (1999-01-08)
A Yeşilkaya, A Yeğin, S Ozdem, T A Aksu
ABSTRACT
Recently, it has been suggested that bilirubin may act as a potent biological chain-breaking antioxidant. To observe the effects of free bilirubin on antioxidant reactions in cumene hydroperoxide-treated erythrocytes (15 g hemoglobin/dl), we added bilirubin at four different concentrations (0.5, 1, 5, and 10 mg/dl). We measured the thiobarbituric acid-reactive substance and reduced glutathione levels, and some antioxidant enzyme activities, namely superoxide dismutase, catalase, and glucose-6-phosphate dehydrogenase. Thiobarbituric acid-reactive substance and chemiluminescent signals decreased during the incubation. Superoxide dismutase activities also decreased but not as much as in the control group. Glucose-6-phosphate dehydrogenase activities and reduced glutathione levels increased, but catalase activities remained the same as the control group. Our results suggest that bilirubin--in the concentrations we have used--partially prevented the oxidant effects of cumene hydroperoxide.
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder