- Pre-embedding immunogold labeling to optimize protein localization at subcellular compartments and membrane microdomains of leukocytes.
Pre-embedding immunogold labeling to optimize protein localization at subcellular compartments and membrane microdomains of leukocytes.
Nature Protocols (2014)
Melo RC