Merck
  • Lactate regulates pyruvate uptake and metabolism in the preimplantation mouse embryo.
Lactate regulates pyruvate uptake and metabolism in the preimplantation mouse embryo.
Biology of reproduction (1999-12-28)
M Lane, D K Gardner
ABSTRACT
This study was an investigation of the interaction of lactate on pyruvate and glucose metabolism in the early mouse embryo. Pyruvate uptake and metabolism by mouse embryos were significantly affected by increasing the lactate concentration in the culture medium. In contrast, glucose uptake was not affected by lactate in the culture medium. At the zygote stage, the percentage of pyruvate taken up and oxidized was significantly reduced in the presence of increasing lactate, while at the blastocyst stage, increasing the lactate concentration increased the percentage of pyruvate oxidized. Lactate oxidation was determined to be 3-fold higher (when lactate was present at 20 mM) at the blastocyst stage compared to the zygote. Analysis of the kinetics of lactate dehydrogenase (LDH) determined that while the V(max) of LDH was higher at the zygote stage, the K(m) of LDH was identical for both stages of development, confirming that the LDH isozyme was the same. Furthermore, the activity of LDH isolated from both stages was reduced by 40% in the presence of 20 mM lactate. The observed differences in lactate metabolism between the zygote and blastocyst must therefore be attributed to in situ regulation of LDH. Activity of isolated LDH was found to be affected by nicotinamide adenine dinucleotide(+) (NAD(+)) concentration. In the presence of increasing concentrations of lactate, zygotes exhibited an increase in autofluorescence consistent with a depletion of NAD(+) in the cytosol. No increase was observed for later-stage embryos. Therefore it is proposed that the differences in pyruvate and lactate metabolism at the different stages of development are due to differences in the in situ regulation of LDH by cytosolic redox potential.
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Pricing and availability is not currently available.