Merck
  • Synergistic effect of HIF-1α and FoxO3a trigger cardiomyocyte apoptosis under hyperglycemic ischemia condition.
Synergistic effect of HIF-1α and FoxO3a trigger cardiomyocyte apoptosis under hyperglycemic ischemia condition.
Journal of cellular physiology (2017-10-17)
Ya-Fang Chen, Sudhir Pandey, Cecilia Hsuan Day, Yu-Feng Chen, Ai-Zhi Jiang, Tsung-Jung Ho, Ray-Jade Chen, Vijaya V Padma, Wei-Wen Kuo, Chih-Yang Huang
ABSTRACT
Cardiomyocyte death is an important pathogenic feature of ischemia and heart failure. Through this study, we showed the synergistic role of HIF-1α and FoxO3a in cardiomyocyte apoptosis subjected to hypoxia plus elevated glucose levels. Using gene specific small interfering RNAs (siRNA), semi-quantitative reverse transcriptase polymerase chain reaction (RT-PCR), Western blot, immunofluorescence, nuclear and cytosolic localization and TUNEL assay techniques, we determined that combined function of HIF-1α and FoxO3a under high glucose plus hypoxia condition lead to enhanced expression of BNIP3 inducing cardiomyocyte death. Our results highlighted the importance of the synergistic role of HIF-1α and FoxO3a in cardiomyocyte death which may add insight into therapeutic approaches to pathophysiology associated with ischemic diabetic cardiomyopathies.
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)