Merck
  • Step-Up Synthesis of Periodic Mesoporous Organosilicas with a Tyrosine Framework and Performance in Horseradish Peroxidase Immobilization.
Step-Up Synthesis of Periodic Mesoporous Organosilicas with a Tyrosine Framework and Performance in Horseradish Peroxidase Immobilization.
Chemistry, an Asian journal (2017-10-19)
Jianqiang Wang, Wenqi Zhang, Changqing Gu, Wenpei Zhang, Man Zhou, Zhiwei Wang, Cheng Guo, Linbing Sun
ABSTRACT
New amino-acid-bridged periodic mesoporous organosilicas (PMOs) were constructed by hydrolysis and condensation reactions under acid conditions in the presence of a template. The tyrosine bissilylated organic precursor (TBOS) was first prepared through a multistep reaction by using tyrosine (a natural amino acid) as the starting material. PMOs with the tyrosine framework (Tyr-PMOs) were constructed by simultaneously using TBOS and tetraethoxysilane as complex silicon sources in the condensation process. All the Tyr-PMOs materials were characterized by XRD, FTIR spectroscopy, N
Sigma-Aldrich
Allyl methyl sulfone, 96%