Merck
Compensatory Internalization of Pma1 in V-ATPase Mutants in
Genetics (2017-12-20)
Swetha Devi Velivela, Patricia M Kane
ABSTRACT
Loss of V-ATPase activity in organelles, whether through V-ATPase inhibition or V-ATPase (
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid