Merck
Low atrazine dosages reduce sperm quality of Calomys laucha mice.
Environmental science and pollution research international (2017-11-18)
Graciela Quintana Saalfeld, Antônio Sergio Varela Junior, Tiane Castro, Fernanda Alves Pereira, Stela Mari Meneghello Gheller, Alessandra Cardoso da Silva, Carine Dahl Corcini, Carlos Eduardo da Rosa, Elton Pinto Colares
ABSTRACT
Reproductive effects caused by the exposure to environmentally relevant dosages of atrazine on wild animals are poorly understood. This study evaluated the effects of three dosages of atrazine on sperm parameters of adult Calomys laucha males. Adult mice were orally exposed to dosages of 0 (water and vehicle control), 0.1, 1, and 10 mg/kg of animal weight for a 21-day period. Following exposure, analyses were performed to determine sperm motility parameters, plasma membrane integrity and fluidity, mitochondrial functionality, acrosome integrity, DNA damage, lipid peroxidation, and production of reactive oxygen species (ROS) in the sperm samples. Total and progressive motility were reduced in all dosages in comparison to control groups. Membrane integrity and mitochondrial functionality of sperm were reduced in all dosages, and the sperm membrane fluidity increased in the higher dosages of atrazine (1 and 10 mg/kg), in comparison with the vehicle control. A decrease in the acrosome integrity was noted at 10 mg/kg of atrazine, compared to the control groups. The integrity of DNA, ROS generation, and lipid peroxidation of sperm showed no significant differences when compared with the control groups. These results suggest that exposure to low dosages of atrazine can affect sperm parameters of Calomys laucha and therefore reduce the reproductive capacity of wild rodent species.
Sigma-Aldrich
Merocyanine 540, Dye content 90 %
Pricing and availability is not currently available.