- The effectiveness of electroporation-based nanocurcumin and curcumin treatments on human breast cancer cells.
The effectiveness of electroporation-based nanocurcumin and curcumin treatments on human breast cancer cells.
Proceedings of ESA annual meeting on electrostatics (2014)
Lin W