[Skip to Content](https://www.sigmaaldrich.com#main-content) [![Merck](https://www.sigmaaldrich.com/static/logos/purple/merck.svg)](https://www.sigmaaldrich.com/IT/en) Products Cart0 ITEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/IT/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/IT/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/IT/en)[Solid Phase Microextraction (SPME)](https://www.sigmaaldrich.com/IT/en/applications/analytical-chemistry/sample-preparation/solid-phase-microextraction)Manual SPME Sampling # Manual SPME Sampling [Learn More About SPME](https://www.sigmaaldrich.com/IT/en/applications/analytical-chemistry/sample-preparation/solid-phase-microextraction) Beginners should consider using a manual SPME fiber and holder. The manual system allows the analyst to optimize the extraction process by evaluating different fibers and extraction conditions in a consistent manner. Begin the extraction process by placing the sample into the vial puck on the hotplate. The vial puck allows users to easily index the vial to begin sample agitation. Insert the holder into the sampling stand, passing the needle through the septum that seals the sample vial. Depress the plunger down to expose the fiber to the sample liquid or headspace above the sample. Start the timing of exposure of the SPME fiber for reproducible extractions. After a predetermined time, the SPME fiber is retracted into the protective needle and transferred to the GC for analysis. Inject the SPME fiber into the GC injection port, where the collected organics are quickly transferred to the GC column by heating the fiber. After the thermal transfer of analytes is completed, the fiber is removed and inserted in the next sample vial for extraction. __Related Articles__ - [Biodiesel](https://www.sigmaaldrich.com/IT/en/technical-documents/technical-article/analytical-chemistry/gas-chromatography/biodiesel) - [Automated Solid Phase Microextraction (SPME) Sampling](https://www.sigmaaldrich.com/IT/en/technical-documents/technical-article/analytical-chemistry/solid-phase-microextraction/automated-spme-sampling) - [Bioanalysis with SPME](https://www.sigmaaldrich.com/IT/en/technical-documents/technical-article/analytical-chemistry/solid-phase-microextraction/bioanalysis-with-spme) - [BioSPME for Plasma Protein Binding Assay: Details of Method Development and Optimization](https://www.sigmaaldrich.com/IT/en/technical-documents/technical-article/analytical-chemistry/solid-phase-microextraction/biospme-plasma-protein-binding-assay) - [Solid Phase Microextraction: Frequently Asked Questions](https://www.sigmaaldrich.com/IT/en/technical-documents/technical-article/analytical-chemistry/solid-phase-microextraction/faq) - [GC/MS/MS Analysis of BPA in Energy Drink](https://www.sigmaaldrich.com/IT/en/technical-documents/technical-article/analytical-chemistry/solid-phase-microextraction/gc-ms-ms-analysis-of-bisphenol-a-bpa-in-a-fruit-flavored-energy-drink-cla010218a) - [Rugged SPME Sampling](https://www.sigmaaldrich.com/IT/en/technical-documents/technical-article/analytical-chemistry/solid-phase-microextraction/nitinol-core-spme-fibers) - [SPME Fiber Performance Tips for Volatiles & Semi-Volatiles](https://www.sigmaaldrich.com/IT/en/technical-documents/technical-article/analytical-chemistry/solid-phase-microextraction/performance-tips) - [View More](https://www.sigmaaldrich.com/IT/en/search/facet-search?focus=sitecontent&term=facet-search) Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/IT/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/IT/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/IT/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/IT/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/IT/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/IT/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/IT/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/IT/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/IT/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/IT/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/IT/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/IT/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/IT/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/IT/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/IT/en/collections/press)[Programs](https://www.sigmaaldrich.com/IT/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/IT/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/merck-life-science) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/Merck_lifesci) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/Merck.lifescience) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/mercklifescience/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@MerckLifeScience/featured) Merck __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/IT/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/IT/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/IT/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/IT/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/IT/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/IT/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/IT/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/IT/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/IT/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/IT/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/IT/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/IT/en/site-map) An unknown error has occured.