Merck
HomeReaction Design & OptimizationAerobic Alcohol Oxidation Solutions
Aerobic Alcohol Oxidation Solutions
Introduction
Alcohol oxidation is one of the most frequently performed oxidation reactions in organic chemistry. The aldehyde and ketone products of alcohol oxidation are useful intermediates en route to complex molecules. Molecular oxygen is an ideal oxidant, but aerobic oxidation reactions are seldom used on a bench scale due to limited synthetic scope, practicality, and safety concerns. The Stahl Aerobic Oxidation Solutions (TEMPO: 796549 and ABNO: 796557), when used with copper source[Cu(MeCN04]OTf (Product No. 685038), operate with ambient air as the source of oxidant and offer a safe and practical way to achieve high reactivity and selectivity in the aerobic oxidation of alcohols to aldehydes and ketones.
Read more about
aerobic alcohol oxidation solution
Advantages
  • Easy reaction set-up (just add substrate and copper catalyst) and work-up procedures
  • Most reactions proceed at room temperature open to the air
  • Reaction completion is indicated by change in color from red/brown to green or blue
  • TEMPO offers chemoselective oxidation of primary alcohols
  • ABNO offers rapid oxidation of both primary and secondary alcohols
  • Broad substrate scope and high functional group tolerance
Representative Applications
aerobic alcohol oxidation solutiontempo and abno
References
1.
Miles K, Stahl S. 2015. Practical Aerobic Alcohol Oxidation with Cu/Nitroxyl and Nitroxyl/NOx Catalyst Systems. Aldrichimica Acta .(48):8-10.
2.
Hoover JM, Stahl SS. 2011. Highly Practical Copper(I)/TEMPO Catalyst System for Chemoselective Aerobic Oxidation of Primary Alcohols. J. Am. Chem. Soc.. 133(42):16901-16910. https://doi.org/10.1021/ja206230h
3.
Hoover JM, Steves JE, Stahl SS. 2012. Copper(I)/TEMPO-catalyzed aerobic oxidation of primary alcohols to aldehydes with ambient air. Nat Protoc. 7(6):1161-1166. https://doi.org/10.1038/nprot.2012.057
4.
Steves JE, Stahl SS. 2013. Copper(I)/ABNO-Catalyzed Aerobic Alcohol Oxidation: Alleviating Steric and Electronic Constraints of Cu/TEMPO Catalyst Systems. J. Am. Chem. Soc.. 135(42):15742-15745. https://doi.org/10.1021/ja409241h
5.
Greene JF, Hoover JM, Mannel DS, Root TW, Stahl SS. 2013. Continuous-Flow Aerobic Oxidation of Primary Alcohols with a Copper(I)/TEMPO Catalyst. Org. Process Res. Dev.. 17(10):1247-1251. https://doi.org/10.1021/op400207f