Merck
  • Chemerin suppresses neuroinflammation and improves neurological recovery via CaMKK2/AMPK/Nrf2 pathway after germinal matrix hemorrhage in neonatal rats.
Chemerin suppresses neuroinflammation and improves neurological recovery via CaMKK2/AMPK/Nrf2 pathway after germinal matrix hemorrhage in neonatal rats.
Brain, behavior, and immunity (2018-03-03)
Yixin Zhang, Ningbo Xu, Yan Ding, Yiting Zhang, Qian Li, Jerry Flores, Mina Haghighiabyaneh, Desislava Doycheva, Jiping Tang, John H Zhang
ABSTRACT
Chemerin, an adipokine, has been reported to reduce the production of pro-inflammatory cytokines and neutrophil infiltration. This study investigated the role of Chemerin and its natural receptor, ChemR23, as well as its downstream mediator calmodulin-dependent protein kinase kinase 2 (CAMKK2)/adenosine monophosphate-activated protein kinase (AMPK) /Nuclear factor erythroid 2-related factor 2 (Nrf2) following germinal matrix hemorrhage (GMH) in neonatal rats, with a specific focus on inflammation. GMH was induced by intraparenchymal injection of bacterial collagenase (0.3U) in P7 rat pups. The results demonstrated that human recombinant Chemerin (rh-Chemerin) improved neurological and morphological outcomes after GMH. Rh-Chemerin promoted accumulation and proliferation of M2 microglia in periventricular regions at 72 h. Rh-Chemerin increased phosphorylation of CAMKK2, AMPK and expression of Nrf2, and decreased IL-1beta, IL-6 and TNF-alpha levels. Selective inhibition of ChemR23/CAMKK2/AMPK signaling in microglia via intracerebroventricular delivery of liposome-encapsulated specific ChemR23 (Lipo-alpha-NETA), CAMKK2 (Lipo-STO-609) and AMPK (Lipo-Dorsomorphin) inhibitor increased the expression levels of IL-1beta, IL-6 and TNF- alpha, demonstrating that ChemR23/CAMKK2/AMPK signaling in microglia suppressed inflammatory response after GMH. Cumulatively, these data showed that rh-Chemerin ameliorated GMH-induced inflammatory response by promoting ChemR23/CAMKK2/AMPK/Nrf2 pathway, and M2 microglia may be a major mediator of this effect. Thus, rh-Chemerin can serve as a potential agent to reduce the inflammatory response following GMH.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human CMKLR1