Merck
Early onset collagen VI myopathies: Genetic and clinical correlations.
Annals of neurology (2010-10-27)
Laura Briñas, Pascale Richard, Susana Quijano-Roy, Corine Gartioux, Céline Ledeuil, Emmanuelle Lacène, Samira Makri, Ana Ferreiro, Svetlana Maugenre, Haluk Topaloglu, Göknur Haliloglu, Isabelle Pénisson-Besnier, Pierre-Yves Jeannet, Luciano Merlini, Carmen Navarro, Annick Toutain, Denys Chaigne, Isabelle Desguerre, Christine de Die-Smulders, Murielle Dunand, Bernard Echenne, Bruno Eymard, Thierry Kuntzer, Kim Maincent, Michèle Mayer, Ghislaine Plessis, François Rivier, Filip Roelens, Tanya Stojkovic, Ana Lía Taratuto, Fabiana Lubieniecki, Soledad Monges, Christine Tranchant, Louis Viollet, Norma B Romero, Brigitte Estournet, Pascale Guicheney, Valérie Allamand
ABSTRACT
Mutations in the genes encoding the extracellular matrix protein collagen VI (ColVI) cause a spectrum of disorders with variable inheritance including Ullrich congenital muscular dystrophy, Bethlem myopathy, and intermediate phenotypes. We extensively characterized, at the clinical, cellular, and molecular levels, 49 patients with onset in the first 2 years of life to investigate genotype-phenotype correlations. Patients were classified into 3 groups: early-severe (18%), moderate-progressive (53%), and mild (29%). ColVI secretion was analyzed in patient-derived skin fibroblasts. Chain-specific transcript levels were quantified by quantitative reverse transcriptase polymerase chain reaction (qRT-PCR), and mutation identification was performed by sequencing of complementary DNA. ColVI secretion was altered in all fibroblast cultures studied. We identified 56 mutations, mostly novel and private. Dominant de novo mutations were detected in 61% of the cases. Importantly, mutations causing premature termination codons (PTCs) or in-frame insertions strikingly destabilized the corresponding transcripts. Homozygous PTC-causing mutations in the triple helix domains led to the most severe phenotypes (ambulation never achieved), whereas dominant de novo in-frame exon skipping and glycine missense mutations were identified in patients of the moderate-progressive group (loss of ambulation). This work emphasizes that the diagnosis of early onset ColVI myopathies is arduous and time-consuming, and demonstrates that quantitative RT-PCR is a helpful tool for the identification of some mutation-bearing genes. Moreover, the clinical classification proposed allowed genotype-phenotype relationships to be explored, and may be useful in the design of future clinical trials.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo anti-collagene di tipo VI, clone VI-26, clone VI-26, Chemicon®, from mouse