Merck
  • ELISA using a recombinant chimera of ESAT-6/MPB70/MPB83 for Mycobacterium bovis diagnosis in naturally infected cattle.
ELISA using a recombinant chimera of ESAT-6/MPB70/MPB83 for Mycobacterium bovis diagnosis in naturally infected cattle.
The Journal of veterinary medical science (2018-10-12)
Ingrid Ieda Fernando de Souza, Rudielle de Arruda Rodrigues, Kláudia Santos Gonçalves Jorge, Márcio Roberto Silva, Walter Lilenbaum, Carlos Eugênio Soto Vidal, Rodrigo Nestor Etges, Miladin Kostovic, Flábio Ribeiro Araújo
ABSTRACT
Bovine tuberculosis (bTB) control programs generally rely on intradermal tuberculin tests for the antemortem diagnosis of Mycobacterium bovis infection in cattle, but these tests detect only a portion of the infected animals. The aim of the present study was to evaluate the diagnostic coverage of a combination of the bTB antemortem techniques known as the comparative intradermal tuberculin test (CITT) and an ELISA based on a recombinant chimera of ESAT-6/MPB70/MPB83 as the antigen in cattle. The results were compared to postmortem findings based on M. bovis culturing and PCR. Paired comparisons of all data (n=92) demonstrated that ELISA and LST results compared to the culturing results did not present significant differences (P=0.27 on McNemar's test and P=0.12 on Fisher's exact test, respectively). Using culturing as the gold standard, the sensitivity and specificity of ELISA were 79.5% (95% CI: 64.5-89.2%) and 75.5% (95% CI: 62.4-85.1%), respectively, whereas LST demonstrated 100% sensitivity (95% CI: 91.03-100%) and 92.5% specificity (95% CI: 82.1-97.0%). The ELISA results did not reveal significant differences in relation to the LST results (P>0.99 on Fisher's exact test). Using the latter as the gold standard, the sensitivity and specificity of ELISA were 79.1% (95% CI: 64.8-88.6%) and 79.6% (95% CI: 66.4-88.5%), respectively. The use of ELISA with the recombinant chimera of ESAT-6/MPB70/MPB83 as the antigen complements the diagnostic coverage provided by CITT and increases the removal of infected animals from herds.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sistema a substrato liquido 3,3′,5,5′-Tetrametil-benzidina (TMB) per ELISA, peroxidase substrate
Sigma-Aldrich
Anti-Bovine IgG (whole molecule)−Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution