Merck
  • Involvement of Gastrin-Releasing Peptide Receptor in the Regulation of Adipocyte Differentiation in 3T3-L1 Cells.
Involvement of Gastrin-Releasing Peptide Receptor in the Regulation of Adipocyte Differentiation in 3T3-L1 Cells.
International journal of molecular sciences (2018-12-14)
Mi-Kyoung Kim, Hyun-Joo Park, Yeon Kim, Soo-Kyung Bae, Hyung Joon Kim, Moon-Kyoung Bae
ABSTRACT
Gastrin-releasing peptide (GRP), a member of bombesin-like peptides, and its receptor (GRP-R) play an important role in various physiological and pathological conditions. In this work, we investigated the role of GRP-R on adipogenesis in 3T3-L1 adipocytes. The expression of GRP-R was significantly increased during the adipocyte differentiation of 3T3-L1 cells. The inhibition of GRP-R by the antagonist RC-3095 affected adipogenesis in 3T3-L1 cells, which reduced lipid accumulation and regulated the expression of adipogenic genes. Moreover, cyclic AMP response element-binding protein (CREB) directly bound to the GRP-R promoter upon exposure to adipogenic stimuli. The down-regulation of GRP-R by the knockdown of CREB inhibited adipocyte differentiation of 3T3-L1 cells. Together these results suggest that the regulation of GRP-R activity or expression has an influence on adipogenesis through regulating adipogenic related genes.