- Transendothelial movement of adiponectin is restricted by glucocorticoids.
Transendothelial movement of adiponectin is restricted by glucocorticoids.
The Journal of endocrinology (2017-07-15)
Thanh Q Dang, Nanyoung Yoon, Helen Chasiotis, Emily C Dunford, Qilong Feng, Pingnian He, Michael C Riddell, Scott P Kelly, Gary Sweeney
PMID28705835
ABSTRACT
Altered permeability of the endothelial barrier in a variety of tissues has implications both in disease pathogenesis and treatment. Glucocorticoids are potent mediators of endothelial permeability, and this forms the basis for their heavily prescribed use as medications to treat ocular disease. However, the effect of glucocorticoids on endothelial barriers elsewhere in the body is less well studied. Here, we investigated glucocorticoid-mediated changes in endothelial flux of Adiponectin (Ad), a hormone with a critical role in diabetes. First, we used monolayers of endothelial cells
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto