All Photos(1)
Z708372
Microcentrifuge tube holder
General description
Snap on tube holders to provide secure lid closure. Large tab facilitates tube removal from centrifuges, water baths, racks and heaters. Polypropylene holders securely fit 0.5 and 1.5 mL standard microcentrifuge tubes. Autoclavable in assorted colors - blue, green, pink, yellow and orange.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).