Skip to Content
Merck

900721

Sigma-Aldrich

Bovine fibrillar collagen solution

type I, 65 mg/mL, pH 7.0, suitable for biomedical research

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

CAS Number:
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

bovine

description

% Denatured: ≤15%
Collagen concentration (by biruret method): 6.0 - 7.0%
Electrophoretic pattern (purity by SDS PAGE): ≤10% collagen faster than alpha.

form

solution

shelf life

24 mo. (from date of manufacture)

concentration

65 mg/mL

impurities

<0.25 EU/mL Endotoxin

pH

7.0-7.6
7.0

storage temp.

2-8°C

General description

Bovine fibrillar collagen solution is a highly concentrated (65 mg/mL) Type I collagen solution that can easily be diluted as needed with media or buffer solutions. This material features high purity (purification through pepsin treatment) which offers reduced antigenicity with the removal of possible contaminants and is provided at neutral pH (pH 7.0). In vivo, fibrillar collagen degrades in approximately 30 days.

Application

Collagen is a major structural protein that provides structure and strength to tissues in the body. Due to its biocompatibility, biodegradability, and high abundance, collagen has been utilized for many applications in biomedical research. Applications of fibrillar collagen may include:
  • Development of collagen-based tissues for skin replacement, bone substitutes, and to generate artificial blood vessels and valves.
  • Drug delivery systems such as sponges for burns/wounds, formulations with liposomes for sustained drug delivery, materials for transdermal drug delivery, and nanoparticles for gene delivery.
  • As collagen scaffolds for research in cancer, electrophysiological protocols, neuron myelination, immunology, and other cell culture applications.

Analysis Note

Appearance: White or off-white viscous gel (paste).

Legal Information

Product of Collagen Solutions.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

900721-VAR:
900721-1EA:
900721-1EA-PW:
900721-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service