Skip to Content
Merck

FLUH9AD3CEF1

Fluorochem (Preferred Partner)

Sodium hydroxide, powder

Sign In to View Organizational & Contract Pricing

Select a Size

500 G

¥12,600

1 KG

¥15,500

Vendor SKU: F996963-500G

¥12,600


Ships in 8 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/Na.H2O/h;1H2/q+1;

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-N

storage temp.

Ambient Storage

Purity

98%

Physical Form

Solid

Product Link

https://fluorochem.co.uk/product/F996963

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service