Merck
All Photos(1)

1.01681

SAFC

Benzonase® ELISA kit II

suitable for detection

Quality Level

application(s)

detection
pharma/biopharma processes

storage temp.

2-8°C

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Benzonase® ELISA kit II can be used for the immunological detection and quantification of the amount of Benzonase® endonuclease present in samples, thus allowing proof of its removal. The use of specific antibodies against Benzonase® endonuclease results in a precise assay with a sensitivity of 0.2 ng/mL. This corresponds to less than 1 ppm in the presence of other proteins (at concentrations higher than 0.5 mg/mL). Cross-reactions with E. coli, Pichia pastoris, normal mouse serum, bovine serum albumin, and fetal calf serum are less than 1%.

Packaging

1 set in EPS box

Analysis Note

Background (AU 450 nm): ≤ 0.2
Linearity: 10 - 1 ng/ml
Detection Limit: 0.2 ng/ml

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service