Merck
All Photos(4)

11741985001

Roche

mRNA Isolation Kit

Synonym(s):
mRNA isolation, nucleic acid isolation

manufacturer/tradename

Roche

Quality Level

packaging

kit of for isolation of at least 70 μg of poly (A)+ RNA

General description

The mRNA Isolation Kit isolates highly purified mRNA from cells or tissue.

Affinity purification is a versatile and highly specific technique for the purification of all classes of biomolecules utilizing differences in biological activities of chemical structures. The high selectivity of this technique results in good purification and high recovery. Often a concentrating effect is reached which enables large volumes to be conveniently processed. The mRNA Isolation Kit depends upon the affinity of the poly(A) + tail of mRNA for a biotin-labeled oligo (dT) probe. The probe can "pull" the mRNA selectively from a lysate without interacting with other RNA or DNA. Once formed, the biotinylated dT-A hybrids can be immobilized on solid surfaces that have been coated with streptavidin, and then washed free of unbound contaminants.

Application

The mRNA Isolation Kit prepares highly purified poly (A) + RNA which may be used directly in many molecular biology applications:

  • RT-PCR
  • cDNA synthesis
  • Northern blotting
  • Northern ELISA
  • RNase protection assay
  • In vitro translation

Features and Benefits

  • Save time. Isolate mRNA directly from cell lysates and tissue homogenates (isolation of total RNA not required).
  • Accommodate a wide range of samples. Work with both small and large scale preparations of mRNA.
  • Achieve highly effective purification. Generate mRNA that is intact and free of DNA and other RNAs.
  • Increase safety. Eliminate the use of hazardous organic solvents.

Components

  • Lysis Buffer
  • Streptavidin-Coated Magnetic Particles
  • Oligo (dT)20 Probe, biotin-labeled
  • Washing Buffer
  • Water, double-distilled, PCR Grade
  • Storage Buffer

Quality

Tested for absence of RNases according to the current quality control procedures; function tested.

Preparation Note

The poly (A)+ tail of mRNA hybridizes to a biotin-labeled oligo(dT) probe. Streptavidin-coated magnetic particles capture the biotinylated hybrids and the particles are separated using a magnetic particle separator. Contaminants are removed by washing, followed by elution of the mRNA from the particles using water.

Analysis Note

Typical Sample*
Cultured cells: 2 x 107
Tissue: 200 mg
Total RNA: 500 μg
* Larger or smaller samples may also be used.

Time Required: Approximately 30 minutes
Product: Poly(A)+ RNA, in double-distilled water

Other Notes

For general laboratory use.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service