Merck
All Photos(1)

93676

Supelco

Elemental Impurities according to USP <2232> dietary supplements

(in 12% nitric acid), TraceCERT®

Quality Level

grade

certified reference material
TraceCERT®

Agency

according to USP

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

As, 15 mg/L
Cd, 5 mg/L
Hg, 15 mg/L
Pb, 5 mg/L

concentration

(in 12% nitric acid)

technique(s)

ICP: suitable

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service