VT000123
certified reference material
ISO 17034
ISO/IEC 17025
solid
limited shelf life, expiry date on the label
package of 10 discs
microbiology
−20°C
Escherichia coli
Escherichia spp.
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | VT000136 | VT000133 | VT000126 |
---|---|---|---|
grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
agency ISO 17034 | agency ISO 17034, ISO/IEC 17025 | agency ISO 17034, ISO/IEC 17025 | agency ISO 17034, ISO/IEC 17025 |
form solid | form disc (hydrosoluble), solid | form disc (hydrosoluble), solid | form disc (hydrosoluble), solid |
shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
packaging package of 10 discs | packaging package of 10 discs | packaging package of 10 discs | packaging package of 10 discs |
application(s) microbiology | application(s) environmental | application(s) environmental | application(s) microbiology |
11 - Combustible Solids
WGK 3
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service