JP
EN
Applications
Products
Services
Support
Home
Search Results
Glycerol, Molecular Biology Grade - CAS 56-81-5 - Calbiochem
(1)
Glycerol, Molecular Biology Grade - CAS 56-81-5 - Calbiochem
Synonyms:
Glycerol, 1,2,3-Propanetriol, Glycerin
Linear Formula:
HOCH
2
CH(OH)CH
2
OH
CAS Number:
56-81-5
Molecular Weight:
92.09
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
356352-M
Glycerol, Molecular Biology Grade - CAS 56-81-5 - Calbiochem
Expand
View Pricing