Merck
Thiamine hydrochloride SAJ special grade, ≥98.5%

Thiamine hydrochloride

Thiamine hydrochloride SAJ special grade, ≥98.5%
Synonyms:
Aneurine hydrochloride, Vitamin B1 hydrochloride
Empirical Formula (Hill Notation):
C12H17ClN4OS · HCl
CAS Number:
Molecular Weight:
337.27

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Thiamine hydrochloride, meets USP testing specifications
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
Thiamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Thiamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Thiamine hydrochloride, ≥98%, FCC, FG
Thiamine hydrochloride, tested according to Ph. Eur.
Thiamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Thiamine hydrochloride, SAJ special grade, ≥98.5%